missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Protector™ Standard Storage Cabinet

The Labconco Protector™ Standard Storage Cabinet may be used for storage of supplies and small laboratory instruments.

Brand:  Labconco™ 9904100

Code : 03

Additional Details : Weight : 40.91000kg

 View more versions of this product

Product Code. 15149715

  • £1129.64 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Specifications

ADA
Single, Right Hinge, Manual Close
80 to 83.18cm
362.8kg
Standard Storage Cabinet
45.72cm
55.88cm
Ideal for storage and venting of small laboratory instrument and mild acid and other reagent
Four Leveling Feet and One 8 in. D Filler Panel to Increase Cabinet Depth from 22 to 30 in.
Epoxy-Coated Steel
38.55kg
Videos