missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Labconco™ Protector™ Standard Storage Cabinet

The Labconco Protector™ Standard Storage Cabinet may be used for storage of supplies and small laboratory instruments.

Brand:  Labconco™ 9900500

Code : 03

Additional Details : Weight : 45.45000kg

 View more versions of this product

Product Code. 15139715

  • £1002.24 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Specifications

55.88cm
Ideal for storage and venting of small laboratory instrument and mild acid and other reagent
Four Leveling Feet and One 8 in. D Filler Panel to Increase Cabinet Depth from 22 to 30 in.
Epoxy-Coated Steel
29.48kg
Single, Right Hinge, Manual Close
90.17 to 93.34cm
362.8kg
Standard Storage Cabinet
30.48cm
Videos