missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Protector™ Standard Storage Cabinet

The Labconco Protector™ Standard Storage Cabinet may be used for storage of supplies and small laboratory instruments.

Brand:  Labconco™ 9900000

Code : 03

Additional Details : Weight : 68.18000kg

 View more versions of this product

Product Code. 15179705

  • £1445.00 / Each


Estimated Shipment
20-07-2018
Add to basket

Includes: Four leveling feet and one 8 in. filler panel to increase cabinet depth from 22 to 30 in. .

Description

Description

  • Support loads up to 800 lb. (363kg)
  • Durable epoxy-coated steel construction
  • Attractive glacier white exterior complements laboratory casework and Labconco fume hoods
  • Manual-closing, non-locking door(s)

Order Info

Work surface and interior shelf kits are sold separately

Specifications

Specifications

White
Ideal for storage and venting of small laboratory instrument and mild acid and other reagent
Four Leveling Feet and One 8 in. D Filler Panel to Increase Cabinet Depth from 22 to 30 in.
Epoxy-Coated Steel
Epoxy
121.92cm
Dual, Manual Close
90.17 to 93.34cm
362.87kg
Standard Storage Cabinet
117.93kg
Videos