Learn More
Labconco™ FlaskScrubber™ Pipet Inserts
Wash and dry pipets inside and out
POA
Specifications
Quantity | Each |
---|---|
For Use With (Application) | Used for direct injection cleaning of pipets |
For Use With (Equipment) | FlaskScrubber™, FlaskScrubber™ Vantage series, SteamScrubber™ glassware washer |
Material | Stainless Steel (304) |
Product Type | Pipet Insert |
Description
Wash and dry pipets inside and out
- Constructed of Type 304 stainless steel
- Self-sealing silicone check valves permit washing of full and partial loads
- Comes with two 1 1/4 in. mounting spindles for attachment to any two locations on the spindle rack (Cat. No. 15-352-828) or with one mounting spindle and attaches to one spindle location (Cat. No. 15-352-808F)
Specifications
Each | |
FlaskScrubber™, FlaskScrubber™ Vantage series, SteamScrubber™ glassware washer | |
Pipet Insert |
Used for direct injection cleaning of pipets | |
Stainless Steel (304) |