Learn More
Labconco™ FlaskScrubber™ Glassware Holders
Glassware Holders for use with Labconco FlaskScrubber Glassware Washers.
Brand:  Labconco™ 4424600
Code : 03
Additional Details : Weight : 1.50000kg
 View more versions of this product
Product Code. 15173103
  • £55.21 / Each
Call for latest availability
+44 (0)1509 555 500
Add to basket
Description
Description
Glassware Holders are made of vinyl-coated Type 304 stainless steel with height adjustment screw and fit onto spindles to grip glassware and hold it in place. For use on upper or lower racks of FlaskScrubber Glassware Washers.
Specifications
Specifications
Vinyl
1 erlenmeyer flask or wide mouth beaker
Used to grip glassware over spindles
Type 304 Stainless Steel
Glassware Holder
Gray
Each
FlaskScrubber™, SteamScrubber™ glassware washer
10.1cm
0.18kg
Videos