Learn More
Labconco™ Drying Chambers for FreeZone™ Freeze-Dry Systems
For use on FreeZone™ Freeze-Dry Systems. Drying Chambers provide a means to attach flasks to your freeze dryer or contain samples in bulk.
Brand: Labconco™ 7522800
Code : 50
Additional Details : Weight : 5.00000kg
Description
For use on FreeZone™ Freeze-Dry Systems: 1, 2.5, 4.5, 6, 8, 12 or 18 Liter models
- Place small sample inside freeze-dry flasks and attach to valves
- Type 304 stainless-steel construction
- Weld-free for dependable vacuum operation
- Neoprene valves accommodate both 0.5 and 0.75 in. adapters, gaskets and clear acrylic top for complete visibility during bulk drying
- Compatible with FreeZone 1, 2.5, 4.5, 6, 8, 12 or 18 Liter Freeze Dryers
Specifications
Drying Chamber | |
12 | |
FreeZone 1, 2.5, 4.5, 6, 8, 12 or 18 Liter Freeze Dryers. | |
8 in. | |
9.75 in. | |
11 lb. |
Stainless Steel | |
To provide a means to attach flasks to freeze dry system or contain samples in bulk | |
12 neoprene valves | |
20 cm | |
24.7 cm | |
5 kg |