missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labconco™ Clear Stoppering Chambers
For use with 6L, 12L and 18L Freezone™ Dryers
£6700.00
Specifications
Type | Clear Stoppering Chamber |
---|---|
For Use With (Application) | To provide economical means of stoppering serum bottles |
For Use With (Equipment) | For 6l, 12l and 18l freezone™ dryers |
Diameter (English) | 13.5 in. |
Diameter (Metric) | 34 cm |
Description
- Allows flasks attachment with freeze dry system or contain samples in bulk
Specifications
Clear Stoppering Chamber | |
For 6l, 12l and 18l freezone™ dryers | |
34 cm | |
58.4 cm | |
30 lb. |
To provide economical means of stoppering serum bottles | |
13.5 in. | |
23 in. | |
50/60 Hz | |
13.6 kg |