missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Clear Drying Chambers for FreeZone™ Freeze-Dry Systems

For visible bulk drying

Brand:  Labconco 7442900

Code : 50

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10235112

  • £1605.88 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Specifications

Clear
For visible bulk drying
33cm
Drying Chamber
4.3kg
26cm
For any freezone™ freeze dry system
Acrylic lid with neoprene gasket, vacuum release valves and pass-through for electrical transformer cords, clear acrylic attachment port lid with 3 in. dia. opening and neoprene gasket
Small
Videos
Documentation