missing translation for 'onlineSavingsMsg'
Learn More

Clear Drying Chambers for FreeZone™ Freeze-Dry Systems

For visible bulk drying

Brand:  Labconco 7443500

Code : 50

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10123342

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

Clear
For visible bulk drying
31cm
Drying Chamber
6.1kg
37cm
For any freezone™ freeze dry system
Acrylic cylinder, anodized aluminum lid, eight neoprene valves with molded plastic knobs, clear acrylic attachment port lid with 3 in. dia. opening and neoprene gasket, valves
Small
Videos
Documentation