missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Clear Drying Chambers for FreeZone™ Freeze-Dry Systems

For visible bulk drying

Brand:  Labconco™ 7444000

Code : 50

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10307092

  • £3113.65 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

  • Durable and corrosion-resistant
  • Chambers without valves include clear acrylic top and bottom lids, gaskets, vacuum break valve, and electrical pass-through for transformer cords
  • Chambers with valves include aluminum lid with eight valves that accommodate both 1/2 in. and 3/4 in. adapters for connecting flasks
  • Use with or without product shelves (shelves available separately, visit www.fishersci.com)

Specifications

Specifications

Clear
For visible bulk drying
38.1cm
Drying Chamber
6.8kg
44cm
For 6, 12, and 18l freezone™ freeze dry systems
Acrylic cylinder, anodized aluminum lid, eight neoprene valves with molded plastic knobs, clear acrylic attachment port lid with 3 in. dia. opening and neoprene gasket, valves
Large
Videos
Documentation