missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Canopy Connections

For Purifier™ Class I and HEPA-filtered Enclosures and XPert™ Filtered Balance Systems

Brand:  Labconco™ 3924401

Code : 03

Additional Details : Weight : 25.00000kg

 View more versions of this product

Product Code. 15199715

  • £539.34 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

Allow Purifier Class I and HEPA-filtered Enclosures and XPert Filtered Balance Systems to be thimble-ducted to the outside for additional protection.

  • Canopy's air gap provides means of balancing air by allowing room air to enter ductwork and mix with filtered air being exhausted from enclosure
  • Outlet: 6.06 in. I.D. and 6.19 in. O.D. and accepts 6 in. nominal diameter ductwork

Specifications

Specifications

15.24cm
Balancing air by allowing room air to enter the ductwork
36 in. Purifier™ class I safety or HEPA filtered enclosure
8.2kg
Videos