missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Canopy Connections

For Purifier™ Class I and HEPA-filtered Enclosures and XPert™ Filtered Balance Systems

Brand:  Labconco™ 3744000

UNSPSC : 41100000

Code : 03

Additional Details : Weight : 2.00000kg

 View more versions of this product

Product Code. 13496708

  • £886.46 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications Description & Specifications

Specifications

15.24cm
Balancing air by allowing room air to enter the ductwork
24 in. Purifier™ class I safety or HEPA filtered enclosure
6.8kg