missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Lab Vision™ Trypsin (Enzymatic Pretreatment) Buffer + 30mL Trypsin Concentrate
Epredia™ Lab Vision Trypsin (Enzymatic Pretreatment) is ideal for immunohistochemical procedures.
£180.00
Specifications
Content And Storage | 2° to 8°C. |
---|---|
Product Type | Trypsin |
Quantity | 1 Ea. |
For Use With (Application) | Immunohistochemistry |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
12623977
|
Epredia™
TA-125-TR |
125mL Buffer + 30mL Trypsin concentrate |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Supplied as a two-component system
Storage Conditions: Store at 2°-8°C. Stable for 18 months.
Includes: TA-015-TR: 15mL (Buffer) (TA-015-TRB), 5mL (Trypsin concentrate) (TA-005-TRC)
TA-125-TR: 125mL (Buffer) (TA-125-TRB), 30mL (Trypsin concentrate) (TA-030-TRC)
Specifications
2° to 8°C. | |
Trypsin | |
1 Ea. | |
Immunohistochemistry |