missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Lab Vision™ IgD (δ-Heavy Chain) Ab-1 Rabbit Polyclonal Antibody
Recommended for Immunohistochemistry (Formalin/paraffin), Epredia™ IgD (δ-Heavy Chain) Ab-1, Rabbit Polyclonal Antibody is one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.
£99.35 - £157.08
Specifications
Antigen | IgD (delta-Heavy Chain) Ab-1 |
---|---|
Clone | Polyclonal |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Host Species | Rabbit |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
12622907
|
Epredia™
RB-1436-A0 |
Concentrated | 100 μL |
£99.35
100µL |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12632907
|
Epredia™
RB-1436-A1 |
Purified | 500 μL |
£146.00
500µL |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12662907
|
Epredia™
RB-1436-R7 |
Prediluted | 7 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10174419
|
Epredia™
RB-1436-A |
Purified | 1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
The most common feature of plasmacytomas, and certain non-Hodgkin's lymphomas is the restricted expression of a single heavy chain class. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: IgD isolated from a pool of normal human sera
Molecular Weight: 50kDa
Positive Control: Tonsil
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
IgD (delta-Heavy Chain) Ab-1 | |
Immunohistochemistry (Paraffin) | |
Rabbit | |
Immunoglobulins | |
IgD isolated from a pool of normal human sera | |
Primary |
Polyclonal | |
Polyclonal | |
IVD | |
Human | |
IgG |