Learn More
Tocris Bioscience™ L189
DNA ligase I, III and IV inhibitor
Brand:  Tocris Bioscience™ 356150
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13433204
  • £515.97 / 50mg
Estimated Shipment date
from Supplier 24-03-2020
Description
Description
DNA ligase I, III and IV inhibitor (IC 50 values are 5, 9 and 5 μM respectively) that blocks DNA binding (K i = 5 μM for DNA ligase I). Preferentially inhibits step 2 of the ligation reaction, and inhibits base excision repair (BER) and non-homologous end joining (NHEJ). Specifically sensitizes cancer cells to DNA damage and increases the cytotoxicity of DNA-damaging agents.
Specifications
Specifications
L189
L189
246.29