missing translation for 'onlineSavingsMsg'
Learn More

L-Tyrosine tert-butyl ester, 99%, Thermo Scientific™

Catalog Number p-4228950
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
16874-12-7
C13H19NO3
237.30
MFCD00042644
DIGHFXIWRPMGSA-LDGXTIHJNA-N
l-tyrosine tert-butyl ester, h-tyr-otbu, tert-butyl l-tyrosinate, l-tyrosine t-butyl ester, tert-butyl 2s-2-amino-3-4-hydroxyphenyl propanoate, h-tyr-obut, tyrosine, 1,1-dimethylethyl ester, 2-amino-3-4-hydroxy-phenyl-propionic acid tert-butyl ester, l-tyrosine tert.butyl ester, pubchem7068
6950582
tert-butyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate
CC(C)(C)OC(=O)[C@@H](N)CC1=CC=C(O)C=C1
This item is not returnable. View return policy
16874-12-7
C13H19NO3
237.30
MFCD00042644
DIGHFXIWRPMGSA-LDGXTIHJNA-N
l-tyrosine tert-butyl ester, h-tyr-otbu, tert-butyl l-tyrosinate, l-tyrosine t-butyl ester, tert-butyl 2s-2-amino-3-4-hydroxyphenyl propanoate, h-tyr-obut, tyrosine, 1,1-dimethylethyl ester, 2-amino-3-4-hydroxy-phenyl-propionic acid tert-butyl ester, l-tyrosine tert.butyl ester, pubchem7068
6950582
tert-butyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate
CC(C)(C)OC(=O)[C@@H](N)CC1=CC=C(O)C=C1

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 142.5°C to 143.8°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Specific Rotation 25
Formula Weight 237.3
Percent Purity 99%
Physical Form Powder
Chemical Name or Material L-Tyrosine tert-butyl ester
Show More Show Less
compliance-icons
Product Identifier
  • L-Tyrosine tert-butyl ester
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'