Learn More
L-Tyrosine tert-butyl ester, 99%, ACROS Organics™
Brand: Acros Organics 435420010
Code : 29
Additional Details : CAS Number : 16874-12-7 Weight : 0.05100kg
Quantity | 1g |
---|---|
Packaging | Glass bottle |
Chemical Identifiers
L-Tyrosine tert-butyl ester | |
99% | |
237.3 |
99% | |
C13H19NO3 |
Specifications
L-Tyrosine tert-butyl ester | |
Authentic | |
99% | |
C13H19NO3 | |
237.3 | |
White | |
1g |
16874-12-7 | |
Powder | |
99% | |
Glass bottle | |
25 | |
142.5° to 143.8°C |