missing translation for 'onlineSavingsMsg'
Learn More

L-Tyrosine benzyl ester p-toluenesulfonate salt, 98%, Thermo Scientific Chemicals

Catalog Number 15408276
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
53587-11-4
C23H25NO6S
443.51
MFCD00038959
PJGVHBLZZQDFFM-RSAXXLAASA-N
h-tyr-obzl.tosoh, l-tyrosine benzyl ester p-toluenesulfonate, l-tyrosine benzyl ester tosylate, h-tyr-obzl.tos, l-tyrosine benzyl ester p-toluenesulfonate salt, o-benzyl-l-tyrosine toluene-p-sulphonate, s-benzyl 2-amino-3-4-hydroxyphenyl propanoate 4-methylbenzenesulfonate, benzyl 2s-2-amino-3-4-hydroxyphenyl propanoate; para-toluene sulfonate, tyr-obzl tosoh, h-tyr-obzl??tos
104540
benzyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate;4-methylbenzenesulfonic acid
CC1=CC=C(C=C1)S(O)(=O)=O.N[C@@H](CC1=CC=C(O)C=C1)C(=O)OCC1=CC=CC=C1
This item is not returnable. View return policy
53587-11-4
C23H25NO6S
443.51
MFCD00038959
PJGVHBLZZQDFFM-RSAXXLAASA-N
h-tyr-obzl.tosoh, l-tyrosine benzyl ester p-toluenesulfonate, l-tyrosine benzyl ester tosylate, h-tyr-obzl.tos, l-tyrosine benzyl ester p-toluenesulfonate salt, o-benzyl-l-tyrosine toluene-p-sulphonate, s-benzyl 2-amino-3-4-hydroxyphenyl propanoate 4-methylbenzenesulfonate, benzyl 2s-2-amino-3-4-hydroxyphenyl propanoate; para-toluene sulfonate, tyr-obzl tosoh, h-tyr-obzl??tos
104540
benzyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate;4-methylbenzenesulfonic acid
CC1=CC=C(C=C1)S(O)(=O)=O.N[C@@H](CC1=CC=C(O)C=C1)C(=O)OCC1=CC=CC=C1

L-Tyrosine benzyl ester p-toluenesulfonate salt, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Tyrosine benzyl ester p-toluenesulfonate salt, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Beilstein 3855339
Solubility Information Slightly soluble in water.
Formula Weight 443.51
Percent Purity 98%
Quantity 25 g
Physical Form Solid
Chemical Name or Material L-Tyrosine benzyl ester p-toluenesulfonate salt

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'