missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ L-Tyrosine benzyl ester p-toluenesulfonate salt, 98%

Brand:  Alfa Aesar™ H63361.22

Code : NEW

Additional Details : CAS Number : 53587-11-4 Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 15418276

  • £150.40 / 100g


Estimated Shipment date
from Supplier 09-07-2018
Add to basket

Description

Description

Applications
L-Tyrosine benzyl ester p-toluenesulfonate salt, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

L-Tyrosine benzyl ester p-toluenesulfonate salt
443.51
3855339
53587-11-4
MFCD00038959
H-Tyr-Obzl.Tos-OH

Specifications

L-Tyrosine benzyl ester p-toluenesulfonate salt
Solid
443.51
3855339
100g
53587-11-4
98%
MFCD00038959
H-Tyr-Obzl.Tos-OH
Videos
SDS