missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ L-Tyrosine benzyl ester, 95%

Brand:  Alfa Aesar™ H64401.06

Code : NEW

Additional Details : CAS Number : 52799-86-7 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15411599

  • £13.60 / 5g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

L-Tyrosine benzyl ester
271.32
2901434
52799-86-7
MFCD00191034
(S)-Benzyl 2-amino-3-(4-hydroxyphenyl)propionate; H-Tyr-obzl

Specifications

L-Tyrosine benzyl ester
Solid
271.32
2901434
94° to 96°C
52799-86-7
95%
MFCD00191034
(S)-Benzyl 2-amino-3-(4-hydroxyphenyl)propionate; H-Tyr-obzl
5g
Videos
SDS