missing translation for 'onlineSavingsMsg'
Learn More

L-Tyrosine, +99%, ACROS Organics™

Brand:  Acros Organics 140640025

UNSPSC : 12350000

Code : 29

Additional Details : CAS Number : 60-18-4 Weight : 2.55000kg

Quantity 2.5kg
Packaging Plastic bottle

 View more versions of this product

Product Code. 10702744

  • £551.90 / 2.50kg


EU Stock 2
Add to basket

Specifications

Chemical Identifiers

L-Tyrosine
99+%
4-(HO)C6H4CH2CH(NH2)CO2H
MFCD00002606
≥99.0% (on dried substance)
C9H11NO3
181.19
3-(4-Hydroxyphenyl)-L-alanine

Specifications

L-Tyrosine
200ppm max.
10ppm max.
10ppm max.
0.5% max. (1 g, 105°C)
99+%
4-(HO)C6H4CH2CH(NH2)CO2H
181.19
14, 605
3-(4-Hydroxyphenyl)-L-alanine
0.1% max.
− 11.05 (20.00°C c=5,dilHCL/H2O 50/50)
2.5kg
60-18-4
400ppm max.
Authentic
Crystalline Powder
≥99.0% (on dried substance)
C9H11NO3
Plastic bottle
MFCD00002606
15, 10020
-9.8° to -12.3° (20°C, 589nm) (c=5, 1N HCl)
300ppm max.
White
Documentation

Technical Documents