missing translation for 'onlineSavingsMsg'
Learn More

L-trans-2,4-PDC, Tocris Bioscience™

Transportable EAAT1-4 inhibitor/non-transportable EAAT5 inhibitor

Brand:  Tocris (Bio-Techne) 0298/10

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13457533

  • £164.64 / 10mg

Estimated Shipment date
from Supplier 06-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent, competitive, transportable EAAT1-4 inhibitor/non-transportable EAAT5 inhibitor. Also available as part of the Excitatory Amino Acid Transporter Inhibitor Tocriset™ .
Specifications

Chemical Identifiers

64769-66-0
159.141
trans-4-carboxy-l-proline, l-trans-pyrrolidine-2,4-dicarboxylic acid, t-pdc, l-trans-2,4-pdc, 2s,4r-pyrrolidine-2,4-dicarboxylic acid, 2,4-pyrrolidinedicarboxylic acid, 2s,4r, tocris-0298, t-2,4-pdc, 2,4-pyrrolidinedicarboxylicacid, 2s,4r
(2S,4R)-pyrrolidine-2,4-dicarboxylic acid
C6H9NO4
NRSBQSJHFYZIPH-DMTCNVIQSA-N
1515192
C1C(CNC1C(=O)O)C(=O)O

Specifications

10mg
64769-66-0
trans-4-carboxy-l-proline, l-trans-pyrrolidine-2,4-dicarboxylic acid, t-pdc, l-trans-2,4-pdc, 2s,4r-pyrrolidine-2,4-dicarboxylic acid, 2,4-pyrrolidinedicarboxylic acid, 2s,4r, tocris-0298, t-2,4-pdc, 2,4-pyrrolidinedicarboxylicacid, 2s,4r
C1C(CNC1C(=O)O)C(=O)O
159.141
159.14
L-trans-2, 4-PDC
C6H9NO4
NRSBQSJHFYZIPH-DMTCNVIQSA-N
(2S,4R)-pyrrolidine-2,4-dicarboxylic acid
1515192
>97%
Videos
Documents