missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Serine ethyl ester hydrochloride, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
L-Serine ethyl ester hydrochloride is widely used in pharmaceutical and food industry.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with oxidizing agents and moisture. Store under dry inert gas. Protect from humidity and water.
L-Serine ethyl ester hydrochloride is widely used in pharmaceutical and food industry.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with oxidizing agents and moisture. Store under dry inert gas. Protect from humidity and water.
Specifications
Melting Point | 130°C to 132°C |
Beilstein | 3562346 |
Solubility Information | Soluble in water. |
IUPAC Name | ethyl (2S)-2-amino-3-hydroxypropanoate;hydrochloride |
Formula Weight | 169.61 |
Percent Purity | 99% |
Quantity | 5 g |
Optical Rotation | −4.4° (c=4 in Water) |
Sensitivity | Hygroscopic |
Chemical Name or Material | L-Serine ethyl ester hydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'