missing translation for 'onlineSavingsMsg'
Learn More

L-Proline tert-butyl ester, 97%, Thermo Scientific Chemicals

Catalog Number p-4228883
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2812-46-6
C9H17NO2
171.24
XJJBXZIKXFOMLP-ZETCQYMHSA-N
l-proline t-butyl ester, h-pro-otbu, tert-butyl l-prolinate, tert-butyl 2s-pyrrolidine-2-carboxylate, l-proline tert-butyl ester, s-tert-butyl pyrrolidine-2-carboxylate, l-proline, 1,1-dimethylethyl ester, l-proline-t-butyl ester, l-proline t-butyl ester free base, t-butyl prolinate
6453108
tert-butyl (2S)-pyrrolidine-2-carboxylate
CC(C)(C)OC(=O)C1CCCN1
This item is not returnable. View return policy
2812-46-6
C9H17NO2
171.24
XJJBXZIKXFOMLP-ZETCQYMHSA-N
l-proline t-butyl ester, h-pro-otbu, tert-butyl l-prolinate, tert-butyl 2s-pyrrolidine-2-carboxylate, l-proline tert-butyl ester, s-tert-butyl pyrrolidine-2-carboxylate, l-proline, 1,1-dimethylethyl ester, l-proline-t-butyl ester, l-proline t-butyl ester free base, t-butyl prolinate
6453108
tert-butyl (2S)-pyrrolidine-2-carboxylate
CC(C)(C)OC(=O)C1CCCN1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Boiling Point 50°C to 51°C (1.0 Torr)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Specific Rotation -40° to -48° (20°C, 589nm) (c=1, ethanol)
Formula Weight 171.24
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material L-Proline tert-butyl ester
Show More Show Less
compliance-icons
Product Identifier
  • L-Proline tert-butyl ester
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'