Learn More
Alfa Aesar™ L-(+)-Penicillamine, 98+%
Brand:  Alfa Aesar™ L13923.MD
Code : NEW
Additional Details : CAS Number : 1113-41-3 Weight : 0.01000kg
Quantity 250mg
 View more versions of this product
Product Code. 11348825
  • £15.90 / 250mg
Estimated Shipment date
from Supplier 28-02-2018
Add to basket
Specifications
Chemical Identifiers
L-(+)-Penicillamine
C5H11NO2S
MFCD00064303
14,7088
1113-41-3
149.21
1722374
3,3-Dimethyl-L-cysteine; 3-Mercapto-L-valine
Specifications
L-(+)-Penicillamine
≥98%
149.21
1722374
3,3-Dimethyl-L-cysteine; 3-Mercapto-L-valine
250mg
+63° (c=1 in 1M NaOH)
1113-41-3
C5H11NO2S
MFCD00064303
14,7088
≈200°C
Unpleasant