missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ L-(+)-Penicillamine, 98+%

Brand:  Alfa Aesar™ L13923.03

Code : NEW

Additional Details : CAS Number : 1113-41-3 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 11338825

  • £23.10 / 1g


Estimated Shipment date
from Supplier 27-02-2018
Add to basket

Specifications

Chemical Identifiers

L-(+)-Penicillamine
C5H11NO2S
MFCD00064303
14,7088
1113-41-3
149.21
1722374
3,3-Dimethyl-L-cysteine; 3-Mercapto-L-valine

Specifications

L-(+)-Penicillamine
≥98%
149.21
1722374
3,3-Dimethyl-L-cysteine; 3-Mercapto-L-valine
1g
+63° (c=1 in 1M NaOH)
1113-41-3
C5H11NO2S
MFCD00064303
14,7088
≈200°C
Unpleasant
Videos
SDS