missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L(+)-Norleucine, ≥99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
50 g
Packaging:
Glass Bottle
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Melting Point | >300.0°C |
Color | White |
Assay Percent Range | 99+% |
Infrared Spectrum | Authentic |
Beilstein | 04, 432 |
Loss on Drying | 1% max. (105°C, 4 hrs) |
Merck Index | 15, 6795 |
Other Amino Acids | (TLC) not detectable |
Packaging | Glass Bottle |
Solubility Information | Solubility in water: 1.6 g/100mL (23 c). |
Mehr anzeigen |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'