missing translation for 'onlineSavingsMsg'
Learn More

L(+)-Norleucine, ≥99%, ACROS Organics™

Brand:  Acros Organics 226040050

Code : 29

Additional Details : CAS Number : 327-57-1 Weight : 0.05500kg

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10522241

  • £80.20 / 5g


EU Stock 2
Add to basket

Specifications

Chemical Identifiers

L(+)-Norleucine
≥99%
C6H13NO2
MFCD00064423
327-57-1
99+%
131.17
L-2-Aminohexanoic acid

Specifications

L(+)-Norleucine
Crystalline Powder
99+%
C6H13NO2
1% max. (105°C, 4 hrs)
04, 432
Glass bottle
L-2-Aminohexanoic acid
Solubility in water: 1.6 g/100mL (23 c).
0275.0°C
White to off-white
5g
327-57-1
≥99%
Authentic
131.17
MFCD00064423
(TLC) not detectable
15, 6795
+ 22.00
0.5% max.
+22° (20°C c=2,6N HCl)
> 300.0°C
Videos
SDS