Learn More
L-Noradrenaline, 98%, Thermo Scientific Chemicals
Description
L-Noradrenaline serves as an adrenergic neurotransmitter. It acts as demethylated precursor of epinephrine (adrenaline). It is also used to treat people having low blood pressure, as a sympathomimetic drug to treat people in vasodilatory shock states such as septic shock and neurogenic shock. Further, it is used in the preparation of1,2,3,4-tetrahydro-4,6,7-isoquinolinetriol hydrochloride. In addition to this, it is used in the treatment of hypotension, depression, schizophrenia and attention hyperactivity disorders.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
L-Noradrenaline serves as an adrenergic neurotransmitter. It acts as demethylated precursor of epinephrine (adrenaline). It is also used to treat people having low blood pressure, as a sympathomimetic drug to treat people in vasodil
Specifications
CAS | 51-41-2 |
Melting Point | ∽220°C (decomposition) |
Molecular Formula | C8H11NO3 |
MDL Number | MFCD00025592 |
Quantity | 1 g |
UN Number | UN2811 |
Beilstein | 4231961 |
Sensitivity | Air and light sensitive |
Merck Index | 14,6695 |
Synonym | norepinephrine, l-noradrenaline, levarterenol, arterenol, noradrenaline, levophed, --norepinephrine, l-norepinephrine, r-noradrenaline, r-norepinephrine |
Show More |
RUO – Research Use Only