Learn More
L-NMMA acetate, Tocris Bioscience™
Non-selective NOS inhibitor
Brand:  Tocris (Bio-Techne) 0771/50
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13437983
  • £251.60 / 50mg
Estimated Shipment
14-09-2020
Description
Description
Competitive, irreversible inhibitor of all three NOS isoforms.
Specifications
Chemical Identifiers
53308-83-1
248.283
tilarginine acetate, l-nmma, l-nmma acetate, ng-monomethyl-l-arginine acetate, nomega-me-l-arg, unii-2fl3530af2, ng-me-l-arg, acoh, tilarginine acetate usan, ng-methyl-l-arginine acetate salt, l-ornithine, n5-imino methylamino methyl-cas
acetic acid;(2S)-2-amino-5-[(N'-methylcarbamimidoyl)amino]pentanoic acid
C9H20N4O4
IKPNWIGTWUZCKM-JEDNCBNOSA-N
135242
CC(=O)O.CN=C(N)NCCCC(C(=O)O)N
Specifications
L-NMMA acetate
C9H20N4O4
IKPNWIGTWUZCKM-JEDNCBNOSA-N
acetic acid;(2S)-2-amino-5-[(N'-methylcarbamimidoyl)amino]pentanoic acid
135242
>98%
53308-83-1
tilarginine acetate, l-nmma, l-nmma acetate, ng-monomethyl-l-arginine acetate, nomega-me-l-arg, unii-2fl3530af2, ng-me-l-arg, acoh, tilarginine acetate usan, ng-methyl-l-arginine acetate salt, l-ornithine, n5-imino methylamino methyl-cas
CC(=O)O.CN=C(N)NCCCC(C(=O)O)N
248.283
248.28