Learn More
L-NIL hydrochloride, Tocris Bioscience™
Selective iNOS inhibitor
Brand:  Tocris (Bio-Techne) 113910
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13488713
  • £128.25 / 10mg
Estimated Shipment
04-10-2018
Add to basket
Description
Description
A selective inhibitor of inducible nitric oxide synthase (IC50 = 3.3 μM).
Specifications
Chemical Identifiers
150403-89-7
223.701
N6-(1-Iminoethyl)-L-lysine hydrochloride
(2S)-2-amino-6-(1-aminoethylideneamino)hexanoic acid;hydrochloride
C8H18ClN3O2
HJYWSATZDBEAOS-FJXQXJEOSA-N
9794509
CC(=NCCCCC(C(=O)O)N)N.Cl
Specifications
L-NIL hydrochloride
>98%
N6-(1-Iminoethyl)-L-lysine hydrochloride
CC(=NCCCCC(C(=O)O)N)N.Cl
223.701
223.7
150403-89-7
C8H18ClN3O2
HJYWSATZDBEAOS-FJXQXJEOSA-N
(2S)-2-amino-6-(1-aminoethylideneamino)hexanoic acid;hydrochloride
9794509