missing translation for 'onlineSavingsMsg'
Learn More

L-Methionine ethyl ester hydrochloride, 99%, ACROS Organics™

Brand:  Acros Organics 205410100

Code : 29

Additional Details : CAS Number : 2899-36-7 Weight : 0.06000kg


Quantity 10g
Packaging Glass bottle

 View more versions of this product

Product Code. 10462591

  • £41.50 / 10g


Estimated Shipment
13-02-2018
Add to basket


Specifications

Chemical Identifiers

L-Methionine ethyl ester hydrochloride
2899-36-7
C7H15NO2S·HCl
213.73
99%
99%
CH3SCH2CH2CH(NH2)CO2C2H5·HCl

Specifications

L-Methionine ethyl ester hydrochloride
2899-36-7
+ 21.00
99%
CH3SCH2CH2CH(NH2)CO2C2H5·HCl
+ 21.00 (25.00°C c=2,EtOH)
10g
99%
Glass bottle
99%
C7H15NO2S·HCl
213.73
82.0° to 92.0°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • L-Methionine ethyl ester hydrochloride, 99%

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

SDS