missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Lysine ethyl ester diisocyanate, 97%, Thermo Scientific Chemicals
54.60 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
Description
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.
Solubility
Sparingly soluble in water (0.19 g/L).
Notes
Moisture Sensitive. Store away form water/moisture. Store away from oxidizing agent.
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.
Solubility
Sparingly soluble in water (0.19 g/L).
Notes
Moisture Sensitive. Store away form water/moisture. Store away from oxidizing agent.
Specifications
Color | Yellow to Brown |
Refractive Index | 1.4572 |
Solubility Information | Sparingly soluble in water (0.19g/L). |
Formula Weight | 226.23 |
Percent Purity | 97% |
Quantity | 5 g |
Sensitivity | Moisture sensitive |
Physical Form | Viscous Liquid |
Chemical Name or Material | L-Lysine ethyl ester diisocyanate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'