missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Leucine-4-nitroanilide, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
It is used to detect/evaluate functionality of amino peptidases. The compound is used in an ELISA assay to detect biological functionality of chicken amino peptidase produced by E. coli. It is also employed to characterize the native IRAP (placental leucine amino peptidase). Leu-pNA, Chromogenic Substrate is documented for use with pure samples as well as cell extracts in vitro.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used to detect/evaluate functionality of amino peptidases. The compound is used in an ELISA assay to detect biological functionality of chicken amino peptidase produced by E. coli. It is also employed to characterize the native IRAP (placental leucine amino peptidase). Leu-pNA, Chromogenic Substrate is documented for use with pure samples as well as cell extracts in vitro.
Solubility
Soluble in DMSO (>100 mM), and methanol (>25 mM).
Notes
Store away from oxidizing agents. Keep the container tightly closed in a well ventilated place.
It is used to detect/evaluate functionality of amino peptidases. The compound is used in an ELISA assay to detect biological functionality of chicken amino peptidase produced by E. coli. It is also employed to characterize the native IRAP (placental leucine amino peptidase). Leu-pNA, Chromogenic Substrate is documented for use with pure samples as well as cell extracts in vitro.
Solubility
Soluble in DMSO (>100 mM), and methanol (>25 mM).
Notes
Store away from oxidizing agents. Keep the container tightly closed in a well ventilated place.
Specifications
Melting Point | 89°C to 90°C |
Beilstein | 2218433 |
Solubility Information | Soluble in DMSO (>100 mM),and methanol (>25 mM). |
Formula Weight | 251.29 |
Percent Purity | 99% |
Quantity | 1 g |
Chemical Name or Material | L-Leucine-4-nitroanilide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'