missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Lactic Acid, Multi-Compendial, J.T. Baker™
684.00 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
500 mL
4 L
19 L
Packaging:
Narrow Mouth Amber Glass Bottle
Poly Pail
Specifications
Formula Weight | 90.08 |
Grade | Multi-Compendial |
Quantity | 4 L |
Flash Point | 112.00°C |
Packaging | Narrow Mouth Amber Glass Bottle |
Specific Gravity | 1L = 1.20kg |
Chemical Name or Material | L-Lactic Acid |
Safety and Handling
Signal Word
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P362-Take off contaminated clothing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'