missing translation for 'onlineSavingsMsg'
Learn More

L-alpha-Hydroxyisocaproic acid, 98%, Thermo Scientific Chemicals

Catalog Number 10581261
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
13748-90-8
C6H12O3
132.16
LVRFTAZAXQPQHI-YFKPBYRVSA-N
l-leucic acid, s-2-hydroxy-4-methylpentanoic acid, 2s-2-hydroxy-4-methylpentanoic acid, l-2-hydroxy-4-methylvaleric acid, s-leucic acid, s-2-hydroxy-4-methylvaleric acid, l-2-hydroxyisocaproic acid, l-alpha-hydroxyisocaproic acid, s---2-hydroxyisocaproic acid, pentanoic acid, 2-hydroxy-4-methyl-, 2s
83697
CHEBI:44510
(2S)-2-hydroxy-4-methylpentanoic acid
CC(C)CC(C(=O)O)O
This item is not returnable. View return policy
13748-90-8
C6H12O3
132.16
LVRFTAZAXQPQHI-YFKPBYRVSA-N
l-leucic acid, s-2-hydroxy-4-methylpentanoic acid, 2s-2-hydroxy-4-methylpentanoic acid, l-2-hydroxy-4-methylvaleric acid, s-leucic acid, s-2-hydroxy-4-methylvaleric acid, l-2-hydroxyisocaproic acid, l-alpha-hydroxyisocaproic acid, s---2-hydroxyisocaproic acid, pentanoic acid, 2-hydroxy-4-methyl-, 2s
83697
CHEBI:44510
(2S)-2-hydroxy-4-methylpentanoic acid
CC(C)CC(C(=O)O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 78°C to 80°C
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula (CH3)2CHCH2CH(OH)COOH
Quantity 1 g
Beilstein 03,336
Formula Weight 132.16
Percent Purity 98%
Chemical Name or Material L-α-Hydroxyisocaproic acid
compliance-icons
Product Identifier
  • L-alpha-Hydroxyisocaproic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'