missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L(+)-Homoarginine hydrochloride, 98%

Catalog Number 15179203
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1483-01-8
C7H16N4O2·HCl
224.69
MFCD00012622
YMKBVNVCKUYUDM-JEDNCBNOSA-N
l-homoarginine hydrochloride, s-2-amino-6-guanidinohexanoic acid hydrochloride, l +-homoarginine hydrochloride, h-homoarg-oh.hcl, l-homoarginine hcl, l-+-homoarginine hydrochloride, h-homoarg-oh, h-dl-hcit-oh, h-homoarg-oh hcl, h-har-oh hcl
2723930
(2S)-2-amino-6-(diaminomethylideneamino)hexanoic acid;hydrochloride
C(CCN=C(N)N)CC(C(=O)O)N.Cl
This item is not returnable. View return policy
1483-01-8
C7H16N4O2·HCl
224.69
MFCD00012622
YMKBVNVCKUYUDM-JEDNCBNOSA-N
l-homoarginine hydrochloride, s-2-amino-6-guanidinohexanoic acid hydrochloride, l +-homoarginine hydrochloride, h-homoarg-oh.hcl, l-homoarginine hcl, l-+-homoarginine hydrochloride, h-homoarg-oh, h-dl-hcit-oh, h-homoarg-oh hcl, h-har-oh hcl
2723930
(2S)-2-amino-6-(diaminomethylideneamino)hexanoic acid;hydrochloride
C(CCN=C(N)N)CC(C(=O)O)N.Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 213.0°C to 215.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula H2NC(=NH)NH(CH2)4CH(NH2)CO2H·HCl
Quantity 25 g
Specific Rotation + 18.00
Solubility Information Solubility in water: 50mg/mL
Formula Weight 224.69
Percent Purity 98%
Physical Form Granular Powder
Chemical Name or Material L(+)-Homoarginine hydrochloride, 99+%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'