missing translation for 'onlineSavingsMsg'
Learn More

L-Histidine, Pure, Fisher Chemical™

Brand:  Fisher Chemical H/0467/48

Code : 21

Additional Details : CAS Number : 71-00-1 Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 10161760

  • £325.00 / 100g

Estimated Shipment: 23-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

71-00-1
155.16
HNDVDQJCIGZPNO-UHFFFAOYNA-N
6274
(2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid
C6H9N3O2
MFCD00064315
l-histidine, histidine, h-his-oh, glyoxaline-5-alanine, anti-rheuma, l---histidine, istidina, s-histidine, l-histidin
CHEBI:15971
NC(CC1=CN=CN1)C(O)=O

Specifications

71-00-1
MFCD00064315
Plastic powder jar
NC(CC1=CN=CN1)C(O)=O
155.16
CHEBI:15971
Solid
282°C
100g
C6H9N3O2
l-histidine, histidine, h-his-oh, glyoxaline-5-alanine, anti-rheuma, l---histidine, istidina, s-histidine, l-histidin
HNDVDQJCIGZPNO-UHFFFAOYNA-N
(2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid
6274
155.16g/mol
White
7.0 to 8.5
Videos
SDS Documents
Certificates
View accessory products