missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Histidine methyl ester dihydrochloride, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
L-Histidine methyl ester dihydrochloride is used to prepare optically pure L-(+)-ergothioneine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
L-Histidine methyl ester dihydrochloride is used to prepare optically pure L-(+)-ergothioneine.
Solubility
Soluble in dimethyl sulfoxide, methanol and water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is hygroscopic in nature. Store under dry inert gas.
L-Histidine methyl ester dihydrochloride is used to prepare optically pure L-(+)-ergothioneine.
Solubility
Soluble in dimethyl sulfoxide, methanol and water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is hygroscopic in nature. Store under dry inert gas.
Specifications
Melting Point | 198°C (decomposition) |
Beilstein | 3572010 |
Solubility Information | Soluble in dimethyl sulfoxide,methanol and water. |
IUPAC Name | methyl 2-amino-3-(1H-imidazol-5-yl)propanoate hydrochloridyl |
Formula Weight | 242.11 |
Percent Purity | ≥98% |
Quantity | 5 g |
Optical Rotation | +10° (c=2 in water) |
Sensitivity | Hygroscopic |
Chemical Name or Material | L-Histidine methyl ester dihydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'