missing translation for 'onlineSavingsMsg'
Learn More

L(-)-Glutathione, oxidized hydrate, 95%, Thermo Scientific Chemicals

Catalog Number p-3736534
45.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
27025-41-8
C20H32N6O12S2
612.63
MFCD00150701
YPZRWBKMTBYPTK-IOKZUGQQNA-N
oxiglutatione, glutathione disulfide, gssg, oxidized glutathione, glutathione disulphide, glutathione-ssg, l-glutathione oxidized, oxigluthione, glutathone disulfide, glutathione oxidized
65359
CHEBI:17858
(2S)-2-amino-5-[[(2R)-3-[[(2R)-2-[[(4S)-4-amino-4-carboxybutanoyl]amino]-3-(carboxymethylamino)-3-oxopropyl]disulfanyl]-1-(carboxymethylamino)-1-oxopropan-2-yl]amino]-5-oxopentanoic acid
N[C@@H](CCC(=O)N[C@@H](CSSC[C@H](NC(=O)CC[C@H](N)C(O)=O)C(=O)NCC(O)=O)C(=O)NCC(O)=O)C(O)=O
This item is not returnable. View return policy
27025-41-8
C20H32N6O12S2
612.63
MFCD00150701
YPZRWBKMTBYPTK-IOKZUGQQNA-N
oxiglutatione, glutathione disulfide, gssg, oxidized glutathione, glutathione disulphide, glutathione-ssg, l-glutathione oxidized, oxigluthione, glutathone disulfide, glutathione oxidized
65359
CHEBI:17858
(2S)-2-amino-5-[[(2R)-3-[[(2R)-2-[[(4S)-4-amino-4-carboxybutanoyl]amino]-3-(carboxymethylamino)-3-oxopropyl]disulfanyl]-1-(carboxymethylamino)-1-oxopropan-2-yl]amino]-5-oxopentanoic acid
N[C@@H](CCC(=O)N[C@@H](CSSC[C@H](NC(=O)CC[C@H](N)C(O)=O)C(=O)NCC(O)=O)C(=O)NCC(O)=O)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 178.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Merck Index 15, 4511
Specific Rotation Condition − 99.00 (20.00°C c=4,water)
Specific Rotation -92° to -106° (20°C, 589nm) (c=2, H2O)
Solubility Information Solubility in water: soluble. Other solubilities: insoluble in most common organic solvents
Formula Weight 612.64
Percent Purity 95%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material L(-)-Glutathione, oxidized hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'