missing translation for 'onlineSavingsMsg'
Learn More

L(-)-Glutathione, oxidized, 98%, Thermo Scientific Chemicals

Catalog Number p-138555
Click to view available options
Quantity:
500 mg
5 g
Packaging:
Glass bottle
27025-41-8
C20H32N6O12S2
612.63
MFCD00150701
YPZRWBKMTBYPTK-IOKZUGQQNA-N
oxiglutatione, glutathione disulfide, gssg, oxidized glutathione, glutathione disulphide, glutathione-ssg, l-glutathione oxidized, oxigluthione, glutathone disulfide, glutathione oxidized
65359
CHEBI:17858
2-amino-4-[(2-{[2-(4-amino-4-carboxybutanamido)-2-[(carboxymethyl)carbamoyl]ethyl]disulfanyl}-1-[(carboxymethyl)carbamoyl]ethyl)carbamoyl]butanoic acid
N[C@@H](CCC(=O)N[C@@H](CSSC[C@H](NC(=O)CC[C@H](N)C(O)=O)C(=O)NCC(O)=O)C(=O)NCC(O)=O)C(O)=O
27025-41-8
C20H32N6O12S2
612.63
MFCD00150701
YPZRWBKMTBYPTK-IOKZUGQQNA-N
oxiglutatione, glutathione disulfide, gssg, oxidized glutathione, glutathione disulphide, glutathione-ssg, l-glutathione oxidized, oxigluthione, glutathone disulfide, glutathione oxidized
65359
CHEBI:17858
2-amino-4-[(2-{[2-(4-amino-4-carboxybutanamido)-2-[(carboxymethyl)carbamoyl]ethyl]disulfanyl}-1-[(carboxymethyl)carbamoyl]ethyl)carbamoyl]butanoic acid
N[C@@H](CCC(=O)N[C@@H](CSSC[C@H](NC(=O)CC[C@H](N)C(O)=O)C(=O)NCC(O)=O)C(=O)NCC(O)=O)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 178.0°C
Color White
Ignition Residue 0.5% max. (800°C)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 500 mg
Beilstein 04, IV, 3168
Merck Index 15, 4511
Specific Rotation Condition −99.00 (20.00°C c=4,water)
Specific Rotation − 99.00
Solubility Information Solubility in water: soluble.
Formula Weight 612.64
Percent Purity 98%
Physical Form Powder
Chemical Name or Material L(-)-Glutathione, oxidized, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'