missing translation for 'onlineSavingsMsg'
Learn More
Learn More
γ-L-Glutamyl-p-nitroanilide, Hydrochloride, Fisher BioReagents
Click to view available options
Quantity:
500 mg
5 g
25 g
Packaging:
Amber Glass
Description
γ-L-Glutamyl-p-Nitroanilide, Hydrochloride is a water soluble form of GPNA suitable for use as substrate for γ-glutamyl transpeptidase.
Specifications
Color | Beige |
Packaging | Amber Glass |
Quantity | 25 g |
Formula Weight | 303.7 |
Physical Form | Powder or Solid |
Chemical Name or Material | γ-L-Glutamyl-p-Nitroanilide, Hydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'