missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ L-Glutamine tert-butyl ester hydrochloride, 98%

Brand:  Alfa Aesar™ H63868.MD

Code : NEW

Additional Details : CAS Number : 39741-62-3 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15498556

  • £18.80 / 250mg


Estimated Shipment date
from Supplier 03-07-2018
Add to basket

Description

Description

Applications
Reactant in the synthesis of rhenium-glutamine conjugates. It is mainly used in food and pharma industry.

Solubility
Soluble 0.3gram in 2ml water clear solution.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acids, heat.
Specifications

Chemical Identifiers

L-Glutamine tert-butyl ester hydrochloride
238.71
H-Gln-OtBu.HCl; (S)-tert-Butyl 2,5-diamino-5-oxopentanoate hydrochloride
39741-62-3
MFCD00039081

Specifications

L-Glutamine tert-butyl ester hydrochloride
Powder
238.71
H-Gln-OtBu.HCl; (S)-tert-Butyl 2,5-diamino-5-oxopentanoate hydrochloride
250mg
39741-62-3
98%
MFCD00039081
White
Videos
SDS