missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Glutamic acid dibenzyl ester hydrochloride, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
L-Glutamic acid dibenzyl ester hydrochloride oxalate used as a intermediate in pharmaceutical and chemical synthesis. Also used in medicine research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
L-Glutamic acid dibenzyl ester hydrochloride oxalate used as a intermediate in pharmaceutical and chemical synthesis. Also used in medicine research.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
L-Glutamic acid dibenzyl ester hydrochloride oxalate used as a intermediate in pharmaceutical and chemical synthesis. Also used in medicine research.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
Color | White |
Molecular Formula | C17H38ClN |
Synonym | tetradecyltrimethylammonium chloride, myristyltrimethylammonium chloride, n,n,n-trimethyltetradecan-1-aminium chloride, trimethyltetradecylammonium chloride, 1-tetradecanaminium, n,n,n-trimethyl-, chloride, n-tetradecyltrimethylammonium chloride, rmv7nju3fx, unii-rmv7nju3fx, trimethyl tetradecyl azanium chloride, tetradecyl trimethyl ammonium chloride |
InChI Key | CEYYIKYYFSTQRU-UHFFFAOYSA-M |
SMILES | CCCCCCCCCCCCCC[N+](C)(C)C.[Cl-] |
IUPAC Name | trimethyl(tetradecyl)azanium;chloride |
Molecular Weight (g/mol) | 291.95 |
PubChem CID | 20708 |
Formula Weight | 363.84 |
Percent Purity | 98% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'