missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L(+)-Glutamic acid, 99%

Catalog Number p-4510306
17.09 GBP valid until 2024-04-30
Use promo code "21741" to get your promotional price.


Click to view available options
Quantity:
100 g
250 g
1 kg
5 kg
Packaging:
Plastic Bottle
Plastic Drum
56-86-0
C5H9NO4
147.13
MFCD00002634
WHUUTDBJXJRKMK-UHFFFAOYNA-N
l-glutamic acid, glutamic acid, 2s-2-aminopentanedioic acid, h-glu-oh, s-2-aminopentanedioic acid, glutaminol, l-glutaminic acid, glutacid, glutamicol, glutamidex
33032
CHEBI:16015
(2S)-2-aminopentanedioic acid
NC(CCC(O)=O)C(O)=O
56-86-0
C5H9NO4
147.13
MFCD00002634
WHUUTDBJXJRKMK-UHFFFAOYNA-N
l-glutamic acid, glutamic acid, 2s-2-aminopentanedioic acid, h-glu-oh, s-2-aminopentanedioic acid, glutaminol, l-glutaminic acid, glutacid, glutamicol, glutamidex
33032
CHEBI:16015
(2S)-2-aminopentanedioic acid
NC(CCC(O)=O)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Ammonium 200ppm max.
Color White
Chlorine (Cl) 200ppm max.
Heavy Metals (as Pb) 10ppm max.
Assay Percent Range 99%
Infrared Spectrum Authentic
Linear Formula HO2CCH2CH2CH(NH2)CO2H
Iron (Fe) 30ppm max.
Loss on Drying 0.5% (105°C, 3 hrs) max.
Other Amino Acids Not detectable
Packaging Plastic Bottle
Specific Rotation +30.5° to +32.5° (20°C, 589nm) (on dried substance) (c=10, 2N HCl)
Sulfate 300ppm max.
Formula Weight 147.13
Percent Purity 98.5 to 100.5% (on dried substance)
Sulfated Ash 0.1% max.
Sublimation Point >200
Specific Rotation Condition +32.00 (20.00°C c=10,2N HCl)
Quantity 100 g
Physical Form Powder
Chemical Name or Material L(+)-Glutamic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'