missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic acid 5-tert-butyl ester, 98%, Thermo Scientific Chemicals

Catalog Number 15469798
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
2419-56-9
C9H17NO4
203.238
MFCD00038580
OIOAKXPMBIZAHL-LURJTMIESA-N
7010573
(2S)-2-amino-5-[(2-methylpropan-2-yl)oxy]-5-oxopentanoic acid
CC(C)(C)OC(=O)CCC(C(=O)O)N
This item is not returnable. View return policy
2419-56-9
C9H17NO4
203.238
MFCD00038580
OIOAKXPMBIZAHL-LURJTMIESA-N
7010573
(2S)-2-amino-5-[(2-methylpropan-2-yl)oxy]-5-oxopentanoic acid
CC(C)(C)OC(=O)CCC(C(=O)O)N

L-Glutamic acid 5-tert-butyl ester is used as a fine chemical intermediate, pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Glutamic acid 5-tert-butyl ester is used as a fine chemical intermediate, pharmaceutical intermediate.

Solubility
Sparingly soluble in water; practically insoluble in ethanol or ether.

Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Beilstein 2049031
Synonym h-glu otbu-oh, l-glutamic acid 5-tert-butyl ester, h-glu o-t-bu-oh, 2s-2-amino-5-tert-butoxy-5-oxopentanoic acid, l-gamma-t-butyl glutamate, l-glutamic acid,5-1,1-dimethylethyl ester, 5-tert-butyl l-glutamate, l-glutamic acid, 5-1,1-dimethylethyl ester, glu otbu, glu tbu
Solubility Information Sparingly soluble in water; practically insoluble in ethanol or ether.
Formula Weight 203.24
Percent Purity 98%
Quantity 1 g
Physical Form Solid
Chemical Name or Material L-Glutamic acid 5-tert-butyl ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'