missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ L-Glutamic acid 1-tert-butyl ester, 97%

Brand:  Alfa Aesar™ H65342.MD

Code : NEW

Additional Details : CAS Number : 45120-30-7 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15452529

  • £14.70 / 250mg


Estimated Shipment date
from Supplier 05-03-2018
Add to basket

Specifications

Chemical Identifiers

L-Glutamic acid 1-tert-butyl ester
203.24
2414793
45120-30-7
MFCD00038562
H-Glu-OtBu; (S)-4-Amino-5-(tert-butoxy)-5-oxopentanoic acid

Specifications

L-Glutamic acid 1-tert-butyl ester
Powder
203.24
2414793
White
250mg
45120-30-7
97%
MFCD00038562
H-Glu-OtBu; (S)-4-Amino-5-(tert-butoxy)-5-oxopentanoic acid
139° to 145°C
Videos
SDS