missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Glutamic acid 1-methyl ester, 98%, Thermo Scientific Chemicals
19.46 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
Description
L-Glutamic acid 1-methyl ester nonessential amino acid that is important to the mammalian central nervous system. It is also a neurotransmitter for cone photoreceptors in the human brain and is used as a treatment for patients who have liver disease accompanied by encephalopathy (a condition termed ‘hepatic coma’).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in water.
Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Soluble in water.
Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Specifications
Melting Point | 145°C (decomposition) |
Color | White |
Molecular Formula | C8H12N2 |
Beilstein | 1707273 |
Synonym | 2,5-dimethyl-1,4-benzenediamine, 4-amino-2,5-dimethylaniline, 2,5-dimethyl-1,4-phenylenediamine, 2,5-dimethyl-p-phenylenediamine, 2,5-diamino-p-xylene, 2,5-dimethylphenylene-1,4-diamine, 2,5-dimethyl-1,4-benzoquinonediamine, ccris 8118, 1,4-dimethylphenylene-2,5-diamine, unii-re400vn2b4 |
Solubility Information | Soluble in water. |
InChI Key | BWAPJIHJXDYDPW-UHFFFAOYSA-N |
SMILES | CC1=CC(=C(C=C1N)C)N |
IUPAC Name | 2,5-dimethylbenzene-1,4-diamine |
Molecular Weight (g/mol) | 136.20 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'