missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L(-)-Fucose, 97%

Catalog Number p-137586
28.73 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
2438-80-4
C6H12O5
164.16
MFCD00135607
SHZGCJCMOBCMKK-DHVFOXMCSA-N
l---fucose, aldehydo-l-fucose, l-galactose, 6-deoxy, 2s,3r,4r,5s-2,3,4,5-tetrahydroxyhexanal, unii-28ryy2iv3f, l---rhodeose, 28ryy2iv3f, l--fucose, l-rhodeose, 3h-fucose
3034656
CHEBI:48204
C[C@@H]1OC(O)[C@@H](O)[C@H](O)[C@@H]1O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
2438-80-4
C6H12O5
164.16
MFCD00135607
SHZGCJCMOBCMKK-DHVFOXMCSA-N
l---fucose, aldehydo-l-fucose, l-galactose, 6-deoxy, 2s,3r,4r,5s-2,3,4,5-tetrahydroxyhexanal, unii-28ryy2iv3f, l---rhodeose, 28ryy2iv3f, l--fucose, l-rhodeose, 3h-fucose
3034656
CHEBI:48204
C[C@@H]1OC(O)[C@@H](O)[C@H](O)[C@@H]1O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 128.0°C to 141.0°C
Color Beige to Gray-Purple
Loss on Drying 0.5% max. (60°C, 4 hrs) (vacuum)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 1 g
Beilstein 31, 78
Merck Index 15, 4308
Specific Rotation Condition - 78.00 (20.00°C c=4,H2O,NH3)
Specific Rotation − 78.00
Solubility Information Solubility in water: soluble in water
IUPAC Name (3S,4R,5S,6S)-6-methyloxane-2,3,4,5-tetrol
Formula Weight 164.16
Percent Purity 97%
Physical Form Powder
Chemical Name or Material L(-)-Fucose, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'