missing translation for 'onlineSavingsMsg'
Learn More

L(-)-Camphorsulfonic acid, 98%, ACROS Organics™

Brand:  Acros Organics 263645000

Code : NEW

Additional Details : CAS Number : 35963-20-3 Weight : 0.50000kg Transport : UN number : 3261 Chem class : 8 Pack group : II


Packaging Glass bottle
Quantity 500g

 View more versions of this product

Product Code. 13468488

  • £256.60 / 500g

Estimated Shipment date
from Supplier 11-03-2020

Please to purchase this item. Need a web account? Register with us today!



Specifications

Chemical Identifiers

35963-20-3
232.3
MIOPJNTWMNEORI-XVKPBYJWSA-N
5771688
[(1S,4R)-7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl]methanesulfonic acid
C10H16O4S
MFCD00064158
l--camphorsulfonic acid, 1r,4s-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonic acid, 1r---camphor-10-sulfonic acid, unii-y6075i4fxe, camphorsulfonic acid,-, l---camphor-10-sulfonic acid, s-camphorsulfonic acid, 1r,4s-7,7-dimethyl-2-oxobicyclo 2.2.1 hept-1-yl methanesulfonic acid, --camphor-10-sulfonic acid
CHEBI:55401
CC1(C2CCC1(C(=O)C2)CS(=O)(=O)O)C

Specifications

L(-)-Camphorsulfonic acid
Glass bottle
White to Beige
500g
98%
MFCD00064158
16,58
Solubility in water: soluble. Other solubilities: slightly soluble in glacial acetic acid and ethyl,acetate,practically insoluble in ether,soluble in chloroform and alcohol
CC1(C2CCC1(C(=O)C2)CS(=O)(=O)O)C
232.3
CHEBI:55401
Crystalline Powder
Authentic
− 22.00
198.0°C
35963-20-3
C10H16O4S
11,316
l--camphorsulfonic acid, 1r,4s-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonic acid, 1r---camphor-10-sulfonic acid, unii-y6075i4fxe, camphorsulfonic acid,-, l---camphor-10-sulfonic acid, s-camphorsulfonic acid, 1r,4s-7,7-dimethyl-2-oxobicyclo 2.2.1 hept-1-yl methanesulfonic acid, --camphor-10-sulfonic acid
MIOPJNTWMNEORI-XVKPBYJWSA-N
[(1S,4R)-7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl]methanesulfonic acid
5771688
232.3
98%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • L(-)-Camphorsulfonic acid

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Documents